missing translation for 'onlineSavingsMsg'
Learn More

USP34 Polyclonal Antibody, Bethyl Laboratories DFS Item

Rabbit Polyclonal Antibody

$344.88 - $344.88

Specifications

Antigen USP34
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501558544 DFS Item
View Documents
Bethyl Laboratories Inc
A300824AT
20 μL
Each for $344.88
Only null left
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.

Ubiquitin hydrolase that can remove conjugated ubiquitin from AXIN1 and AXIN2, thereby acting as a regulator of Wnt signaling pathway. Acts as an activator of the Wnt signaling pathway downstream of the beta-catenin destruction complex by deubiquitinating and stabilizing AXIN1 and AXIN2, leading to promote nuclear accumulation of AXIN1 and AXIN2 and positively regulate beta-catenin (CTNBB1)-mediated transcription. Recognizes and hydrolyzes the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications

Specifications

USP34
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
deubiquitinating enzyme 34, KIAA0570, KIAA0729, ubiquitin specific protease 34, ubiquitin thioesterase 34, ubiquitin thiolesterase 34, ubiquitin-specific-processing protease 34
Between 3350 and C-term
Primary
4° C
USP34
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse
Q6ZQ93, Q70CQ2
17847, 9736
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents