missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP34 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5104200

Catalog No. PIPA5104200


Only null left

Description

Description

Antibody detects endogenous levels of total USP34.

Ubiquitin hydrolase that can remove conjugated ubiquitin from AXIN1 and AXIN2, thereby acting as a regulator of Wnt signaling pathway. Acts as an activator of the Wnt signaling pathway downstream of the beta-catenin destruction complex by deubiquitinating and stabilizing AXIN1 and AXIN2, leading to promote nuclear accumulation of AXIN1 and AXIN2 and positively regulate beta-catenin (CTNBB1)-mediated transcription. Recognizes and hydrolyzes the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications

Chemical Identifiers

USP34
-20°C
Polyclonal
Liquid
IgG
Human, Mouse
USP34
A530081C03Rik; deubiquitinating enzyme 34; KIAA0570; KIAA0729; Murr2; U2af1-rs1 region 2; ubiquitin carboxyl-terminal hydrolase 34; ubiquitin specific peptidase 34; ubiquitin specific protease 34; ubiquitin thioesterase 34; ubiquitin thiolesterase 34; ubiquitin-specific-processing protease 34; Usp34
USP34
Primary
Affinity Chromatography
1 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6ZQ93, Q70CQ2
17847, 9736
A synthesized peptide derived from human USP34(Accession Q70CQ2), corresponding to amino acid residues S516-D566.
Antibody

Specifications

USP34
Polyclonal
Unconjugated
USP34
A530081C03Rik; deubiquitinating enzyme 34; KIAA0570; KIAA0729; Murr2; U2af1-rs1 region 2; ubiquitin carboxyl-terminal hydrolase 34; ubiquitin specific peptidase 34; ubiquitin specific protease 34; ubiquitin thioesterase 34; ubiquitin thiolesterase 34; ubiquitin-specific-processing protease 34; Usp34
Rabbit
Affinity Chromatography
RUO
17847, 9736
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6ZQ93, Q70CQ2
USP34
A synthesized peptide derived from human USP34(Accession Q70CQ2), corresponding to amino acid residues S516-D566.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions