missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560690 Supplier Invitrogen™ Supplier No. PA560690
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: VVIEPSSVLN GGKYSFGTAA HPMEQVEDRI GSSLSYVNTT EEKFSDNIST ASEASETAGS GFLYSATPGA DVCFARQHNT SDNNNQCLLG ANGNILLHLN PQKPGAIDNQ PLVTQEPVKA TSLTLEGGRL KRTPQLIHGR DYEMVPE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

USP32 (Ubiquitin Specific Peptidase 32) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include calcium ion binding and thiol-dependent ubiquitin-specific protease activity. An important paralog of this gene is USP6. [GeneCards].

Specifications

Antigen USP32
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Usp32
Gene Accession No. Q8NFA0
Gene Alias 2900074J03Rik; 3.4.19.12; 6430526O11Rik; AW045245; Deubiquitinating enzyme 32; NY-REN-60; renal carcinoma antigen NY-REN-60; Ubiquitin carboxyl-terminal hydrolase 32; ubiquitin specific peptidase 32; ubiquitin specific protease 32; ubiquitin thioesterase 32; ubiquitin thiolesterase 32; ubiquitin-specific-processing protease 32; USP10; USP32
Gene Symbols Usp32
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP32. Recombinant protein control fragment (Product #RP-93399).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less