missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP22 Recombinant Rabbit Monoclonal Antibody (18E3)
GREENER_CHOICE

Catalog No. FEMA550121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550121 Supplier Invitrogen™ Supplier No. MA550121

Rabbit Recombinant Monoclonal Antibody

USP22 Recombinant Monoclonal Antibody for ICC/IF, Flow, ELISA

Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.

Specifications

Antigen USP22
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 18E3
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP22
Gene Accession No. Q9UPT9
Gene Alias AI427806; Deubiquitinating enzyme 22; KIAA1063; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific peptidase 22; ubiquitin specific protease 22; ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; ubiquitin-specific-processing protease 22; Usp22; USP3L
Gene Symbols USP22
Host Species Rabbit
Immunogen A synthesized peptide derived from Human USP22.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23326
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less