missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USF1 Recombinant Rabbit Monoclonal Antibody (JE63-68)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA562887
Description
Positive Control: HEK-293 cell lysate, K-562 cell lysate, NIH/3T3 cell lysate, Mouse kidney tissue lysate, Rat kidney tissue lysate, HeLa, mouse bladder tissue, rat bladder tissue.Tissue Specificity: Low tissue specificitySubcellular Location: Nucleus.Sequence Similarities: 99% MousePredicted band size: 34 kDa.
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| USF1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P22415, Q61069 | |
| USF1 | |
| Synthetic peptide within Human USF1 aa 150-250. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE63-68 | |
| Unconjugated | |
| USF1 | |
| bHLHb11; Class B basic helix-loop-helix protein 11; FCHL; FCHL1; HYPLIP1; Major late transcription factor 1; MLTF; MLTFI; UEF; Upstream stimulatory factor 1; upstream transcription factor 1; USF; USF1; usf1-bd | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22278, 7391, 83586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |