missing translation for 'onlineSavingsMsg'
Learn More

Sartorius USB Data Cable
GREENER_CHOICE

Catalog No. 14558847
Change view
Click to view available options
Connection Type:
USB-C to USB-A
USB-C to USB-B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
14558847 USB-C to USB-B
14558848 USB-C to USB-A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14558847 Supplier Sartorius Supplier No. YCCUSBCB

USB Data Cable for Entris™ Lab Balance.

  • To connect Sartorius laboratory balances of the product family Entris™ to a printer or a second display via USB interface
  • Color: Black

Specifications

For Use With (Equipment) Weights and Weight Sets
Connection Type USB-C to USB-B
Length (English) 5 ft.
Length (Metric) 1.5 m
Type Data Cable