missing translation for 'onlineSavingsMsg'
Learn More

Ursolic acid, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015504
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015504 25 mg
111015503 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015504 Supplier Merck Emd Millipore Supplier No. 8979725MGF
Only null left

Ursolic acid belongs to the class of triterpenoid compounds that occurs widely in medicinal herbs, food and other plants. It may possess pharmacological properties like hepta-protection, analgesic, cardiotonic, sedative and tonic effects.

Food and Beverages

Specifications

CAS 77-52-1
Physical Form Neat
Linear Formula C30H48O3
MDL Number MFCD00009621
Melting Point 292°C
Synonym 3 beta-Hydroxy-12-ursen-28-ic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 456.70
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less