missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ursolic acid, ≥98.5% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| CAS | 77-52-1 |
| Physical Form | Neat |
| Linear Formula | C30H48O3 |
| MDL Number | MFCD00009621 |
| Melting Point | 292°C |
| Synonym | 3 beta-Hydroxy-12-ursen-28-ic acid |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 456.70 |
| Percent Purity | ≥98.5% (HPLC) |
| Show More |