missing translation for 'onlineSavingsMsg'
Learn More

Ursodiol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012907
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012907 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012907 Supplier Merck Emd Millipore Supplier No. PHR1579500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ursodiol is an important bile acid, which can be biologically derived from the epimerization of chenodiol by intestinal bacteria. It is widely used in combination with chenodiol for gall stone dissolution and cholestatic liver diseases.

Specifications

CAS 128-13-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H40O4
MDL Number MFCD00003680
Melting Point 203°C to 204°C
Synonym Ursodeoxycholic acid; 3 alpha,7 beta-Dihydroxy-5 beta-cholan-24-oic acid; 5 beta-Cholan-24-oic acid-3 alpha,7 beta-diol; 7 beta-Hydroxylithocholic acid; UDCS
Type Certified Reference Material
Quantity 500 mg
Formula Weight 392.57
Grade Certified Reference Material