missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UROD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104404
Description
Antibody detects endogenous levels of total UROD.
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.Chemical Identifiers
| UROD | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Urod | |
| AI323803; PCT; porphyrinogen carboxylase; porphyrinogen carboxy-lyase; UPD; URO D; UROD; Uro-d; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase | |
| Urod | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P06132, P32362, P70697 | |
| 22275, 29421, 7389 | |
| A synthesized peptide derived from human UROD(Accession P06132), corresponding to amino acid residues E316-Q366. | |
| Antibody |
Specifications
| UROD | |
| Polyclonal | |
| Unconjugated | |
| Urod | |
| AI323803; PCT; porphyrinogen carboxylase; porphyrinogen carboxy-lyase; UPD; URO D; UROD; Uro-d; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22275, 29421, 7389 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P06132, P32362, P70697 | |
| Urod | |
| A synthesized peptide derived from human UROD(Accession P06132), corresponding to amino acid residues E316-Q366. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |