missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uric acid, 99% max., MP Biomedicals™
$228.18 - $228.18
Chemical Identifiers
| CAS | 69-93-2 |
|---|---|
| Molecular Formula | C5H4N4O3 |
| Molecular Weight (g/mol) | 168.112 |
| InChI Key | LEHOTFFKMJEONL-UHFFFAOYSA-N |
| Synonym | uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro |
| PubChem CID | 1175 |
| ChEBI | CHEBI:17775 |
| IUPAC Name | 7,9-dihydro-3H-purine-2,6,8-trione |
| SMILES | C12=C(NC(=O)N1)NC(=O)NC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10321580
|
MP Biomedicals Inc
0210321580 |
100 g |
Each for $228.18
|
|
|||||
Chemical Identifiers
| 69-93-2 | |
| 168.112 | |
| uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro | |
| CHEBI:17775 | |
| C12=C(NC(=O)N1)NC(=O)NC2=O |
| C5H4N4O3 | |
| LEHOTFFKMJEONL-UHFFFAOYSA-N | |
| 1175 | |
| 7,9-dihydro-3H-purine-2,6,8-trione |
Specifications
| 69-93-2 | |
| 1.89g/mL | |
| C5H4N4O3 | |
| uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro | |
| C12=C(NC(=O)N1)NC(=O)NC2=O | |
| 168.112 | |
| CHEBI:17775 | |
| Crystalline Powder |
| White | |
| 99% max. | |
| 100 g | |
| LEHOTFFKMJEONL-UHFFFAOYSA-N | |
| 7,9-dihydro-3H-purine-2,6,8-trione | |
| 1175 | |
| 168.11 | |
| Uric acid |