accessibility menu, dialog, popup

UserName

CAS RN 69-93-2

ONHOHNHNOHN

CAS RN 69-93-2

IUPAC Name: 2,3,6,7,8,9-hexahydro-1H-purine-2,6,8-trione
Synonyms: uric acid acid, uric urate (substance) ua - uric acid uric acid (substance) ammonium acid urate sodium acid urate monohydrate lithic acid sodium acid urate lithate
Molecular Weight (g/mol): 168.11
Molecular Formula: C5H4N4O3
InChi Key: LEHOTFFKMJEONL-UHFFFAOYSA-N
SMILES: O=C1NC2=C(N1)C(=O)NC(=O)N2
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results