missing translation for 'onlineSavingsMsg'
Learn More

Urethane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018658
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018658 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018658 Supplier Merck Emd Millipore Supplier No. 76607100MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 51-79-6
Vapor Pressure 10 mmHg (77.8°C)
Vapor Density 3.07 (vs. air)
Flash Point 92°C - Closed Cup
Physical Form Neat
Linear Formula NH2 COOC2H5
MDL Number MFCD00007966
Melting Point 46°C to 49°C; 48°C to 50°C
Boiling Point 182°C to 184°C
Synonym Carbamic acid ethyl ester; Ethyl carbamate; Ethylurethane
Type Analytical Standard
Quantity 100 mg
Formula Weight 89.09
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less