missing translation for 'onlineSavingsMsg'
Learn More

Urea Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014692
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014692 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014692 Supplier Merck Emd Millipore Supplier No. PHR14061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Specifications

CAS 57-13-6
Density 1.335 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula NH2 CONH2
MDL Number MFCD00008022
Melting Point 132°C to 135°C
Synonym Urea; Carbamide; Carbonyldiamide
Type Certified Reference Material
Quantity 1 g
Formula Weight 60.06
Grade Certified Reference Material
Show More Show Less