missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea Peroxide, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Melting Point | 88° to 93°C |
| Packaging | Amber Glass Bottle |
| Quantity | 100 g |
| Grade | Reagent |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.