missing translation for 'onlineSavingsMsg'
Learn More

Urea hydrogen peroxide adduct, 97%

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol

Supplier:  Thermo Scientific Chemicals L1394022

 View more versions of this product

Catalog No. AAL1394022


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

Solubility
Soluble in water.

Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.
Specifications

Specifications

Urea hydrogen peroxide adduct
∽90°C (decomposition)
CH6N2O3
MFCD00013119
UN1511
Very Hygroscopic
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
AQLJVWUFPCUVLO-UHFFFAOYSA-N
hydrogen peroxide;urea
31294
94.07
124-43-6
Odorless
H2NCONH2·H2O2
100 g
3680414
14,1782
Soluble in water.
C(=O)(N)N.OO
94.07
CHEBI:75178
97%

Chemical Identifiers

124-43-6
94.07
AQLJVWUFPCUVLO-UHFFFAOYSA-N
31294
hydrogen peroxide;urea
CH6N2O3
MFCD00013119
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
CHEBI:75178
C(=O)(N)N.OO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: UREA HYDROGEN PEROXIDE

EINECSNumber : 204-701-4

RTECSNumber : T4860000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only