missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99%, (White crystalline powder or pellets/Ultra Pure), MP Biomedicals

Supplier:  MP Biomedicals Inc 0210569501

 View more versions of this product

Catalog No. ICN10569501

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Urea
White
4X Crystallized
1 lb.
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
1176
60.1
Crystalline Powder or Pellets
57-13-6
99%
CH4N2O
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
C(=O)(N)N
60.056
CHEBI:48376
Ultrapure

Chemical Identifiers

57-13-6
60.056
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
CHEBI:48376
C(=O)(N)N
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
urea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions