missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99%, (White crystalline powder or pellets/Ultra Pure), MP Biomedicals

Catalog No. p-3547380
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10569501 1 lb.
ICN10569505 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10569501 Supplier MP Biomedicals Inc Supplier No. 0210569501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Assay Percent Range 99%
Quantity 1 lb.
Formula Weight 60.1
Grade Ultrapure
Physical Form Crystalline Powder or Pellets
Assay 4X Crystallized
Chemical Name or Material Urea
Recommended Storage Room Temperature