missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, 99% Pure, Promega™
A protein denaturant qualified for use in polyacrylamide gels.
Supplier: Promega Corporation V3171
Specifications
| Urea | |
| 57-13-6 | |
| CH4N2O | |
| carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
| C(=O)(N)N | |
| 60.056 | |
| CHEBI:48376 |
| For use as the denaturing component in polyacrylamide gels | |
| Colorless to White | |
| 1 kg | |
| XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
| urea | |
| 1176 | |
| Crystalline Powder or Free-flowing Crystals |
Chemical Identifiers
| 57-13-6 | |
| 60.056 | |
| carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
| CHEBI:48376 | |
| C(=O)(N)N |
| CH4N2O | |
| XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
| 1176 | |
| urea |
Safety and Handling
Recommended Storage : Store at 22° to 25°C.