missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, 99% Pure, Promega™
A protein denaturant qualified for use in polyacrylamide gels.
$121.21 - $387.37
Chemical Identifiers
CAS | 57-13-6 |
---|---|
Molecular Formula | CH4N2O |
Molecular Weight (g/mol) | 60.056 |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
Synonym | carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate |
PubChem CID | 1176 |
ChEBI | CHEBI:48376 |
IUPAC Name | urea |
SMILES | C(=O)(N)N |
Chemical Identifiers
57-13-6 | |
60.056 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
CHEBI:48376 | |
C(=O)(N)N |
CH4N2O | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
Specifications
57-13-6 | |
CH4N2O | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
C(=O)(N)N | |
60.056 | |
CHEBI:48376 | |
Urea, For use as the denaturing component in polyacrylamide gels |
Colorless to White | |
1 kg | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
Crystalline Powder or Free-flowing Crystals |
Safety and Handling
Recommended Storage : Store at 22° to 25°C.