missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Urea, ≥99%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Manufacturer: Mp Biomedicals 0219145001
Specifications
Urea | |
≥99% | |
alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
ACS Reagent |
57-13-6 | |
CH4N2O | |
1kg | |
C(=O)(N)N | |
60.056 | |
CHEBI:48376 | |
Crystalline |
Chemical Identifiers
57-13-6 | |
60.056 | |
alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert | |
CHEBI:48376 | |
C(=O)(N)N |
CH4N2O | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
Safety and Handling
Recommended Storage : Room Temperature