missing translation for 'onlineSavingsMsg'
Learn More

Urea, ≥99%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Catalog No. p-3547379
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19145001 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19145001 Supplier MP Biomedicals Inc Supplier No. 0219145001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Assay Percent Range ≥99%
Quantity 1 kg
Formula Weight 60.06
Grade ACS Reagent
Physical Form Crystalline
Chemical Name or Material Urea
Recommended Storage Room Temperature