missing translation for 'onlineSavingsMsg'
Learn More

Urea, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014691
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014691 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014691 Supplier Merck Emd Millipore Supplier No. 561801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 57-13-6
Density 1.335 g/mL (at 25°C)
Physical Form Neat
Linear Formula NH2 CONH2
MDL Number MFCD00008022
Melting Point 132°C to 135°C
Synonym Carbamide; Carbonyldiamide
Solubility Completely soluble in water
Type Analytical Standard
Quantity 1 g
Formula Weight 60.06
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less