missing translation for 'onlineSavingsMsg'
Learn More

Urapidil 98.0+%, TCI America™

Catalog No. U00851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00851G Supplier TCI America Supplier No. U00851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34661-75-1
Molecular Formula C20H29N5O3
Molecular Weight (g/mol) 387.484
MDL Number MFCD00133908
InChI Key ICMGLRUYEQNHPF-UHFFFAOYSA-N
Synonym 6-[[3-[4-(2-Methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyluracil
PubChem CID 5639
IUPAC Name 6-[3-[4-(2-methoxyphenyl)piperazin-1-yl]propylamino]-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C(=CC(=O)N(C1=O)C)NCCCN2CCN(CC2)C3=CC=CC=C3OC

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 387.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Urapidil
RTECSNumber YQ9862000
TSCA No