accessibility menu, dialog, popup

UserName

CAS RN 34661-75-1

IUPAC Name: 6-({3-[4-(2-methoxyphenyl)piperazin-1-yl]propyl}amino)-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: urapidil eupressyl ebrantil uraprene urapidil, 5-(14)C-labeled urapidilum urapidil, 2,3-(14)c2-labeled mediatensyl 2,3-(14)C2-labeled urapidil 2,3-(14)c2-labeled urapidil
Molecular Weight (g/mol): 387.48
Molecular Formula: C20H29N5O3
InChi Key: ICMGLRUYEQNHPF-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1N1CCN(CCCNC2=CC(=O)N(C)C(=O)N2C)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results