missing translation for 'onlineSavingsMsg'
Learn More

Uracil Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015110
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015110 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015110 Supplier Merck Emd Millipore Supplier No. PHR1581100MG
Pharmaceutical (small molecule)

Specifications

CAS 66-22-8
Linear Formula C4H4N2O2
MDL Number MFCD00006016
Melting Point >300°C
Synonym Uracil; 2,4(1 H,3 H)-Pyrimidinedione; 2,4-Dihydroxypyrimidine; 2,4-Pyrimidinediol
Type Certified Reference Material
Quantity 100 mg
Formula Weight 112.09
Grade Certified Reference Material