missing translation for 'onlineSavingsMsg'
Learn More

Uracil-5-carboxaldehyde, 97%

Catalog No. p-7033901
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51098MD 250 mg
AAH5109803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51098MD Supplier Thermo Scientific Chemicals Supplier No. H51098MD

CAS: 1195-08-0 | C5H4N2O3 | 140.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1195-08-0
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00192185
InChI Key OHAMXGZMZZWRCA-UHFFFAOYSA-N
Synonym 5-formyluracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, uracil 5-carbaldehyde, uracil-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 1,2,3,4-tetrahydro-2,4-dioxo, 2,4 1h,3h-pyrimidinedione, 5-formyl, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde, 2,3h-pyrimidinedione, 5-formyl, 5-pyrimidinecarboxaldehyde,2,3,4-tetrahydro-2,4-dioxo, zlchem 870
PubChem CID 304590
ChEBI CHEBI:80961
IUPAC Name 2,4-dioxo-1H-pyrimidine-5-carbaldehyde
SMILES O=CC1=CNC(=O)NC1=O

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 140.1
Percent Purity 97%
Chemical Name or Material Uracil-5-carboxaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only