accessibility menu, dialog, popup

UserName

CAS RN 1195-08-0

IUPAC Name: 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde
Synonyms: 5-formyluracil
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: OHAMXGZMZZWRCA-UHFFFAOYSA-N
SMILES: O=CC1=CNC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results