missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149852
Description
UPK2 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes one of the proteins of the highly conserved urothelium-specific integral membrane proteins of the asymmetric unit membrane which forms urothelium apical plaques in mammals. The asymmetric unit membrane is believed to strengthen the urothelium by preventing cell rupture during bladder distention. The encoded protein is expressed in the peripheral blood of bladder cancer patients with transitional cell carcinomas.Specifications
| UPK2 | |
| Polyclonal | |
| Unconjugated | |
| UPK2 | |
| AW491716; UP2; UPII; UPK2; uroplakin 2; uroplakin II; Uroplakin-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 689093, 7379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O00526 | |
| UPK2 | |
| Recombinant Uroplakin 2 (UPK2) (Asp26-Thr153). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |