missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18609841
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609841 50 kg Amber Glass Bottle
18609836 25 g Amber Glass Bottle
18609837 125 g Amber Glass Bottle
18609838 500 g Amber Glass Bottle
18609839 2.5 kg Amber Glass Bottle
18609840 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609841 Supplier Spectrum Chemical Mfg Cor Supplier No. C130550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 50 kg
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Cobalt Chloride, Hexahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.