missing translation for 'onlineSavingsMsg'
Learn More

Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID

Catalog No. 29008954
Encompass
Change view
Click to view available options
Overall Length (Metric):
5 cm
10 cm
20 cm
30 cm
0.5 m
1 m
1.5 m
5 m
Diameter (Metric) Inner:
0.125 mm
0.175 mm
0.25 mm
0.5 mm
0.75 mm
1.0 mm
1.15 mm
2.0 mm
Material:
Stainless Steel (304)
Stainless Steel (316)
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric) Diameter (Metric) Inner Material
29008954 1 m 2.0 mm Stainless Steel (316)
05701106 5 cm 0.75 mm Stainless Steel (316)
05701108 20 cm 0.75 mm Stainless Steel (316)
05701109 30 cm 0.75 mm Stainless Steel (316)
05701119 20 cm 0.175 mm Stainless Steel (316)
05701126 1 m 0.5 mm Stainless Steel (304)
05701127 0.5 m 0.75 mm Stainless Steel (316)
05701128 1 m 0.75 mm Stainless Steel (316)
05701129 5 cm 1.0 mm Stainless Steel (316)
05701130 10 cm 1.0 mm Stainless Steel (316)
05701138 20 cm 1.15 mm Stainless Steel (316)
0570194 5 cm 0.5 mm Stainless Steel (304)
0570195 10 cm 0.5 mm Stainless Steel (304)
0570196 20 cm 0.5 mm Stainless Steel (304)
0570197 30 cm 0.5 mm Stainless Steel (304)
05701141 1 m 1.15 mm Stainless Steel (316)
05701179 5 m 2.0 mm Stainless Steel (316)
05701104 20 cm 0.25 mm Stainless Steel (316)
05701102 5 cm 0.25 mm Stainless Steel (316)
05701100 1.5 m 0.75 mm Stainless Steel (316)
05701117 5 cm 0.175 mm Stainless Steel (316)
05701118 10 cm 0.175 mm Stainless Steel (316)
05701120 30 cm 0.175 mm Stainless Steel (316)
05701124 1 m 0.25 mm Stainless Steel (316)
05701121 0.5 m 0.175 mm Stainless Steel (316)
05701147 0.5 m 0.125 mm Stainless Steel (316)
0570199 1.5 m 0.25 mm Stainless Steel (316)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Catalog No. 29008954 Supplier Idex Supplier No. UP U 800

missing translation for 'validMainframeMsg'

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Type Tubing
Diameter (English) Inner 1/8 in.
Diameter (English) Outer 0.080 in.
Diameter (Metric) Inner 2.0 mm
Overall Length (English) 3.2 ft.
Overall Length (Metric) 1 m
Material Stainless Steel (316)
Maximum Pressure 7600 psi (524 bar)