missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID
Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses
Supplier: Idex U150DUP1
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
- Precut 316 stainless steel tubing:
- Ends are machine cut and polished
- Edges are deburred inside and outside
- Tubing is passivated
- Inside is flushed with reagent-grade isopropanol
- Color-coded banding for easy identification of the inner diameter
- All stainless-steel tubing longer than 1m is coiled
Specifications
| Tubing | |
| 0.046 in. | |
| 3.2 ft. | |
| Stainless Steel (316) |
| 1/16 in. | |
| 1.15 mm | |
| 1 m | |
| 5800 psi (400 bar) |