Learn More
Idex Quick Connect Adapter
Convenience of quick connect and disconnect of a luer connection
Supplier: Idex UPP678
Description
Convenience of quick connect and disconnect of a luer connection
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.Specifications
| Female Luer Adapter | |
| 125°C | |
| Red |
| 0.05 in. | |
| ETFE | |
| Pair to Female 1/4-28 |