Learn More
Idex Quick Connect Adapter DFS Item
Convenience of quick connect and disconnect of a luer connection
Supplier:  Idex UPP675
 View more versions of this product
Catalog No. 05-700-364
Only null left
Explore available promotions
Description
Description
Convenience of quick connect and disconnect of a luer connection
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Specifications
Specifications
Male Luer Adapter
40°C
Red
0.05 in.
ETFE
Male Luer Lock x Female 1/4-28 Flat Bottom
Product Suggestions
Product Suggestions