Learn More
Idex National Pipe Thread (NPT) Adapters DFS Item
Maintain secure connections to 1/8 in. and 1/4 in. NPT ports even under high pressure
Supplier:  Idex UPU510
 View more versions of this product
Catalog No. 29-008-456
Only null left
Explore available promotions
Description
Description
  • Securely join together fluid pathway components
  • Manufactured from PEEK polymer
  • Includes fittings
Specifications
Specifications
Adapter
0.062 in.
P-300/308 for 0.125 in. O.D. tubing
Red
0.125 in. Male NPT to 0.25-28 Female
PEEK
Product Suggestions
Product Suggestions