Learn More
Idex LuerTight™ Fittings: Tefzel DFS Item
Convenience of quick connect and disconnect of a luer connection
$12.81 - $14.94
Specifications
Temperature (Metric) Max 80°C
Material ETFE
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (English) Inner Color For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Color For Use With (Equipment) Price Quantity  
05-700-153 DFS Item
View Documents
Idex
UPP624DUP
0.05 in. Natural To Male 1/4-28
Each for $12.81
Only null left
05-700-158 DFS Item
View Documents
Idex
UPP629
0.04 in. Natural To Female 10-32
Each for $13.77
Only null left
05-701-328 DFS Item
View Documents
Idex
UPP686
0.05 in. Black To Female M6
Each for $12.81
Only null left
05-700-157 DFS Item
View Documents
Idex
UPP628DUP
0.04 in. Natural Female Luer x Female 1/4-28 Flat Bottom
Each for $14.94
Only null left
Description
Description
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
80°C
ETFE
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'