Learn More
Idex Low Pressure Y Connectors
Combine, split, or intersect fluid streams throughout your system
Supplier:  Idex UP P512
Encompass
 View more versions of this product
Catalog No. 05701369
Only null left
Description
Description
Combine, split, or intersect fluid streams throughout your system
  • Manufactured from inert polymers
Use Y connectors in a variety of low-pressure situations throughout your fluidic system. Y connectors create less turbulence and solvent outgassing than Tee connectors, improving analytical results. Handy mounting holes make it a snap to mount connectors. Y connector assemblies come complete with nuts and ferrules.
Specifications
Chemical Identifiers
Low Pressure Y Connector
1000 psi (69 bar)
PEEK
Split a Stream or Join Two Streams Together
Specifications
Low Pressure Y Connector
1000 psi (69 bar)
P-235/P-200 fittings
Split a Stream or Join Two Streams Together
0.02 in.
17 μL
PEEK
Product Suggestions
Product Suggestions