Learn More
Upchurch Scientific™ Inline Check Valve Cartridge
Catalog No. 05704198
Encompass
Change view
Click to view available options
For Use With (Equipment):
CV-3000
Catalog No. For Use With (Equipment)
05704198 CV-3000
1 options
Metal fittings are not recommended with the use of any inline check valve. Use of the improper fittings may damage the check valves and render them non-functional.
Catalog No. 05704198 Supplier Upchurch Scientific™ Supplier No. UP CV 3001
Only null left
Metal fittings are not recommended with the use of any inline check valve. Use of the improper fittings may damage the check valves and render them non-functional.
Limit fluid flow to one direction.
  • Low cracking pressure for easy use in most applications
  • Standard and non-metallic versions for assured chemical compatibility
Specifications
For Use With (Equipment) CV-3000
For Use With (Application) Limit fluid flow to one direction
Max. Pressure 1.5-3 psi Cracking Pressure
Type Inline Check Valve Cartridge
Volume (Metric) Working Less Than 150 uL