Learn More
MilliporeSigma™ Supelco™ Untreated Fused Silica Tubing
Catalog No. 111004926
Change view
Click to view available options
Overall Length (Metric):
5 m
15 m
60 m
Diameter (Metric) Inner:
0.10 mm
0.25 mm
0.32 mm
0.53 mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric) Diameter (Metric) Inner
111004926 5 m 0.10 mm
111004508 15 m 0.25 mm
111003740 60 m 0.25 mm
111005002 5 m 0.32 mm
111003670 60 m 0.32 mm
111004834 5 m 0.53 mm
111004274 15 m 0.53 mm
111003456 60 m 0.53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 111004926 Supplier MilliporeSigma™ Supelco™ Supplier No. 25735
Specifications
Type Tubing
Description Untreated
For Use With (Application) Gas Chromatography (GC)
Diameter (Metric) Inner 0.10 mm
Overall Length (Metric) 5 m
Material Fused Silica