missing translation for 'onlineSavingsMsg'
Learn More

Chiron Unsaturated Telomeric Acid, FTUCAs, and Unsaturated Acid Standards

Catalog No. p-200087331
Change view
Click to view available options
Chemical Name or Material:
2H-Perfluoro-2-decenoic acid
2H-Perfluoro-2-dodecenoic acid
2H-Perfluoro-2-octenoic acid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
141731050ME 2H-Perfluoro-2-decenoic acid
144741250ME 2H-Perfluoro-2-dodecenoic acid
15264850ME 2H-Perfluoro-2-octenoic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 141731050ME Supplier Chiron Supplier No. 14173.1050ME

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material 2H-Perfluoro-2-decenoic acid
CAS 70887-84-2
Concentration 50 μg/mL
Synonym 8:2 FTUCA
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL