missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Shake multiple tubes at an adjustable angle to maximize sample aeration. The rack holders are adjustable to fit any of the four available test tube rack sizes.
$190.13 - $1443.13
Specifications
| For Use With (Equipment) | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
|---|
| Catalog Number | Mfr. No. | Product Type | Tube Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Tube Size | Price | Quantity | ||||
|
11676033
|
Thermo Scientific™
600074 |
Test Tube Rack | 21 to 25 mm |
Each for $290.40
|
|
||||
|
11676034
|
Thermo Scientific™
600075 |
Test Tube Rack | 26 to 30 mm |
Each for $308.28
|
|
||||
|
11676039
|
Thermo Scientific™
600088 |
Test Tube Rack Holder | Varies, Adjustable |
Each for $1,443.13
|
|
||||
|
11676044
|
Thermo Scientific™
950040 |
Test Tube Rack | 10 to 13 mm |
Each for $211.77
|
|
||||
|
11676043
|
Thermo Scientific™
950060 |
Test Tube Rack | 16 to 20 mm |
Each for $190.13
|
|
||||
Description
Shake multiple tubes at an adjustable angle to maximize sample aeration. Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide ultimate flexibility—they can be adjusted to fit any of the four available test tube rack sizes.
- Universal test tube rack holder can be adjusted to fit any of the four available test tube rack sizes
- Universal design requires the separate purchase of a test tube rack
Specifications
| MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |