Learn More
Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Shake multiple tubes at an adjustable angle to maximize sample aeration. The rack holders are adjustable to fit any of the four available test tube rack sizes.
Supplier:  Thermo Scientific™ 950040
 View more versions of this product
Catalog No. 11676044
Only null left
Explore available promotions
Description
Description
Shake multiple tubes at an adjustable angle to maximize sample aeration. Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide ultimate flexibility—they can be adjusted to fit any of the four available test tube rack sizes.
  • Universal test tube rack holder can be adjusted to fit any of the four available test tube rack sizes
  • Universal design requires the separate purchase of a test tube rack
Specifications
Specifications
Tube
3 to 7 mL
MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
10 to 13 mm
Test Tube Rack
72
Product Suggestions
Product Suggestions