missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Universal Plus Absorbent Booms

Universal absorbent booms are ideal for containment both indoors and out.

Supplier:  Brady™ SPCUN510

Catalog No. 17-111-093

Only null left
Explore available promotions

Description

Description

  • Universal Plus booms can be used on land and indoors to absorb all water-based, oil-based and chemical fluids
  • Universal absorbency - picks up water-based, petroleum-based and non-aggressive, chemical fluids
  • Booms are lint-free and easy to handle
  • Included clips allow for connection to extend overall length
Specifications

Specifications

EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
Absorbent Boom
121.2 L
Absorbent Boom, Oil-only
10 ft.
5 in.
Green
32 gal.
Polypropylene
4/Pk.
3.05 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions