missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Globe Scientific Universal Plug Stoppers
Description
- Made from polyethylene
- Available in different colors
Specifications
Specifications
| Product Type | Universal Plug Stopper |
| Closure Size | Universal |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Lavender |
| Sterility | Non-sterile |
| For Use With (Equipment) | 10, 12, 13, 16 mm Tubes |