missing translation for 'onlineSavingsMsg'
Learn More

Labconco Universal Hydraulic Base Stand

Universal Hydraulic Base Stands offer the flexibility to adjust the work surface infinitely from 28 to 40 inches. Programmable features include four memory positions as well as upper and lower travel stop positions.

Supplier:  Labconco 5181300

 View more versions of this product

Catalog No. 17200213


Only null left

Description

Description

  • Support loads up to 1,000 lbs. (454 kg)
  • Durable epoxy-coated Optic White steel frame
  • Working heighs of 28 to 40 in.
  • Four user-programmable memory positions
  • Front-accessible controls that are right or left side compatible
  • Programmable safety stops to limit upper and lower travel ranges
  • Fixed Leveling Feet
Specifications

Chemical Identifiers

Enclosures
Hydraulic Base Stand

Specifications

Enclosures
Hydraulic Base Stand
3 ft.
Casters
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions