Learn More
Corning™ Universal Fit Hinged Rack Pipet Tips, Sterile
Hinged closure provides easy stacking and steam penetration during autoclaving
Supplier: Corning™ C4711
Description
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
| 1 to 200 μL | |
| Yellow | |
| Hinged Rack | |
| Hinged rack |
| Sterile | |
| Hinged closure provides easy stacking and steam penetration during autoclaving | |
| Round | |
| Yes |