missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Universal 7 Pipette Work Station, Model 337

Catalog No. 02912675
Change view
Click to view available options
Color:
Ice Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02912675 Ice Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02912675 Supplier DWK Life Sciences™ Supplier No. W851354

Keep your workstation organized with this universal pipette stand for up to 7 pipettes. Available in a variety of colors.

  • Space-saving pipette stand holds up to seven single-channel pipettes of most brands
  • Made of sturdy polyamid material, easy to clean
  • Heavy bottom plate and anti-slip pads guarantee bench stability
  • The work station fits new and old generations of Socorex and other brand pipettes

Specifications

Description Model 337
Color Ice Blue
Holds Pipettes (7 Single Channel)
Material Polyamide
Positions 7
Shape Round
For Use With (Equipment) Univeral
No. of Racks 1 Each
Includes Anti-slip pads
Product Type Pipette Work Station