missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific Supplies Equal Volume Cylinders
Ideal for specific gravity, density, volume and calorimetry experiments.
$51.54
Specifications
| For Use With (Application) | Specific gravity, density, volume and calorimetry experiments |
|---|
| Catalog Number | Mfr. No. | Product Type | Material | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Material | Price | Quantity & Availability | ||||
|
S07034
|
United Scientific Supplies
SGVC04 |
Cylinder Set | Aluminum, Copper, Steel, Brass |
Each for $51.54
|
|
||||
Description
- Available as individual and set of cylinders
- Made of aluminum, brass, copper, steel, PVC and wood
Specifications
| Specific gravity, density, volume and calorimetry experiments |