Learn More
United Scientific Supplies Equal Volume Cylinders
Catalog No. S07034
Change view
Click to view available options
Product Type:
Cylinder Set
Material:
Aluminum, Copper, Steel, Brass
Catalog No. Product Type Material
S07034 Cylinder Set Aluminum, Copper, Steel, Brass
1 options
Catalog No. S07034 Supplier United Scientific Supplies Supplier No. SGVC04
Ideal for specific gravity, density, volume and calorimetry experiments.
  • Available as individual and set of cylinders
  • Made of aluminum, brass, copper, steel, PVC and wood
Specifications
For Use With (Application) Specific gravity, density, volume and calorimetry experiments
Product Type Cylinder Set
Includes Four identically sized cylinders of different metals; Storage box with a fitted compartment
Material Aluminum, Copper, Steel, Brass