missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ UniStore™ Graduated Wide Mouth Bottle

High quality translucent polypropylene with easy to read mL volume increments

Supplier:  United Scientific™ 33700

 View more versions of this product

Catalog No. S01470


Only null left

Description

Description

  • These high-quality translucent polypropylene wide mouth bottles include easy to read mL volume increments
  • The graduation lines are screen printed for sharp contrast, and each bottle has a white write-on area for identification
  • Bottles are autoclavable and are ideal for storing liquids or solids
  • Easy to fill wide mouth design
  • Screen printed volume graduations for sharp contrast
  • Write-on area for labeling
  • Autoclavable at 121°C
  • Leakproof
Specifications

Specifications

125 mL
Translucent
Non-sterile
Yes
Reagent Bottle
Autoclavable
4 oz.
Polypropylene
Yes
Wide, Threaded
Blank
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions